siRNA Details
Gene Symbol: Olfr900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146874.1 | NP_667085.1 | 1 | 521 | ||
GenBank mRNA | BC106986.2 | - | 1 | 521 | 40043276 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146874.1 | NP_667085.1 | 1 | 521 | ||
GenBank mRNA | BC106986.2 | - | 1 | 521 | 40043276 |