siRNA Details
| Gene Symbol: Olfr900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146874.1 | NP_667085.1 | 1 | 627 | ||
| GenBank mRNA | BC106986.2 | - | 1 | 627 | 40043276 | |
| Gene Symbol: Olfr898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146871.2 | NP_667082.2 | 1 | 627 | ||
| GenBank mRNA | BC127995.1 | - | 1 | 631 | 40135758 | |

Olfr900
Genomic Map