siRNA Details
| Gene Symbol: Olfr218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001809.2 | NP_001001809.2 | 1 | 311 | ||
| GenBank mRNA | BC104284.1 | - | 1 | 310 | ||
| BC104291.1 | - | 1 | 311 | 40042773 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr218| Gene Symbol: Olfr218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001809.2 | NP_001001809.2 | 1 | 311 | ||
| GenBank mRNA | BC104284.1 | - | 1 | 310 | ||
| BC104291.1 | - | 1 | 311 | 40042773 | ||