siRNA Details
Gene Symbol: Olfr1404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146881.2 | NP_667092.1 | 1 | 666 | ||
GenBank mRNA | BC120856.1 | - | 1 | 666 | 40129779 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146881.2 | NP_667092.1 | 1 | 666 | ||
GenBank mRNA | BC120856.1 | - | 1 | 666 | 40129779 |