siRNA Details
| Gene Symbol: Olfr1294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146885.1 | NP_667096.1 | 1 | 664 | ||
| GenBank mRNA | BC127976.1 | - | 1 | 668 | 40135176 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1294| Gene Symbol: Olfr1294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146885.1 | NP_667096.1 | 1 | 664 | ||
| GenBank mRNA | BC127976.1 | - | 1 | 668 | 40135176 | |