siRNA Details
| Gene Symbol: Olfr1225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146891.2 | NP_667102.2 | 1 | 766 | ||
| GenBank mRNA | BC145849.1 | - | 1 | 766 | 40131152 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1225| Gene Symbol: Olfr1225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146891.2 | NP_667102.2 | 1 | 766 | ||
| GenBank mRNA | BC145849.1 | - | 1 | 766 | 40131152 | |