siRNA Details
| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005160.3 | NP_005151.2 | 17, 18 | 1646 | ||
| XM_011529975.2 | XP_011528277.1 | 16, 17 | 1356 | |||
| GenBank mRNA | AK058103.1 | - | 16, 17 | 1436 | ||
| AK292234.1 | - | 17, 18 | 1512 | |||
| BC036797.1 | - | 19, 20 | 1740 | 5590378 | ||
| X69117.1 | - | 17, 18 | 1499 | |||

GRK3
Genomic Map