siRNA Details
| Gene Symbol: Olfr1205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146896.3 | NP_667107.2 | 2 | 276 | ||
| GenBank mRNA | CB173441.1 | - | 2 | 276 | ||
| Gene Symbol: Olfr1201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146895.1 | NP_667106.1 | 1 | 133 | ||

Olfr1205
Genomic Map