siRNA Details
| Gene Symbol: Olfr1205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146896.3 | NP_667107.2 | 2 | 654 | ||
| GenBank mRNA | CB173413.1 | - | 1 | 344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1205| Gene Symbol: Olfr1205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146896.3 | NP_667107.2 | 2 | 654 | ||
| GenBank mRNA | CB173413.1 | - | 1 | 344 | ||