siRNA Details
| Gene Symbol: Olfr1220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146900.2 | NP_667111.2 | 1 | 581 | ||
| GenBank mRNA | BC125363.1 | - | 1 | 581 | 40129878 | |
| BC131900.1 | - | 1 | 581 | 40130177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1220| Gene Symbol: Olfr1220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146900.2 | NP_667111.2 | 1 | 581 | ||
| GenBank mRNA | BC125363.1 | - | 1 | 581 | 40129878 | |
| BC131900.1 | - | 1 | 581 | 40130177 | ||