siRNA Details
| Gene Symbol: Olfr1217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146901.2 | NP_667112.1 | 1 | 336 | ||
| GenBank mRNA | BC132097.1 | - | 1 | 336 | 40130374 | |
| BC138391.1 | - | 1 | 336 | 8861411 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1217| Gene Symbol: Olfr1217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146901.2 | NP_667112.1 | 1 | 336 | ||
| GenBank mRNA | BC132097.1 | - | 1 | 336 | 40130374 | |
| BC138391.1 | - | 1 | 336 | 8861411 | ||