siRNA Details
| Gene Symbol: Olfr1378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146910.1 | NP_667121.1 | 1 | 881 | ||
| GenBank mRNA | BC106990.2 | - | 1 | 880 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1378| Gene Symbol: Olfr1378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146910.1 | NP_667121.1 | 1 | 881 | ||
| GenBank mRNA | BC106990.2 | - | 1 | 880 | ||