siRNA Details
| Gene Symbol: Olfr1377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146911.1 | NP_667122.1 | 1 | 622 | ||
| GenBank mRNA | BC137754.1 | - | 1 | 667 | 8860773 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1377| Gene Symbol: Olfr1377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146911.1 | NP_667122.1 | 1 | 622 | ||
| GenBank mRNA | BC137754.1 | - | 1 | 667 | 8860773 | |