siRNA Details
| Gene Symbol: Olfr1336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146915.1 | NP_667126.1 | 1 | 879 | ||
| GenBank mRNA | BC104285.1 | - | 1 | 878 | ||
| BC104286.1 | - | 1 | 879 | 40042538 | ||
More Information
Set Membership: |
|

Olfr1336
Genomic Map