siRNA Details
| Gene Symbol: Olfr1346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146916.1 | NP_667127.1 | 1 | 138 | ||
| GenBank mRNA | BC137876.1 | - | 1 | 162 | 8860895 | |
| BC137877.1 | - | 1 | 162 | 8860896 | ||
More Information
Set Membership: |
|

Olfr1346
Genomic Map