siRNA Details
| Gene Symbol: Olfr1180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146918.2 | NP_667129.2 | 1 | 691 | ||
| GenBank mRNA | BC120811.1 | - | 1 | 691 | 40129734 | |
| BC120813.1 | - | 1 | 691 | 40129736 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1180| Gene Symbol: Olfr1180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146918.2 | NP_667129.2 | 1 | 691 | ||
| GenBank mRNA | BC120811.1 | - | 1 | 691 | 40129734 | |
| BC120813.1 | - | 1 | 691 | 40129736 | ||