siRNA Details
| Gene Symbol: Olfr1188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146919.2 | NP_667130.2 | 1 | 489 | ||
| GenBank mRNA | BC104076.1 | - | 1 | 469 | 40041756 | |
| BC104077.1 | - | 1 | 469 | 40041757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1188| Gene Symbol: Olfr1188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146919.2 | NP_667130.2 | 1 | 489 | ||
| GenBank mRNA | BC104076.1 | - | 1 | 469 | 40041756 | |
| BC104077.1 | - | 1 | 469 | 40041757 | ||