siRNA Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006742.2 | NP_006733.1 | 2 | 923 | ||
| GenBank mRNA | AJ272212.1 | - | 2 | 883 | ||
| AK309272.1 | - | 2 | 923 | |||
| BC062616.1 | - | 2 | 894 | 5188431 | ||
| M14504.1 | - | 1 | 159 | |||

PSKH1
Genomic Map