siRNA Details
| Gene Symbol: Olfr476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146924.1 | NP_667135.1 | 1 | 137 | ||
| Gene Symbol: Olfr477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146926.1 | NP_667137.1 | 1 | 137 | ||

Olfr476
Genomic Map