siRNA Details
| Gene Symbol: Olfr807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146929.1 | NP_667140.1 | 1 | 500 | ||
| GenBank mRNA | BC138074.1 | - | 1 | 537 | 8861093 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr807| Gene Symbol: Olfr807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146929.1 | NP_667140.1 | 1 | 500 | ||
| GenBank mRNA | BC138074.1 | - | 1 | 537 | 8861093 | |