siRNA Details
| Gene Symbol: Olfr790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146933.1 | NP_667144.1 | 1 | 263 | ||
| GenBank mRNA | BC125495.1 | - | 1 | 336 | 40130010 | |
| BC132182.1 | - | 1 | 336 | 40130459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr790| Gene Symbol: Olfr790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146933.1 | NP_667144.1 | 1 | 263 | ||
| GenBank mRNA | BC125495.1 | - | 1 | 336 | 40130010 | |
| BC132182.1 | - | 1 | 336 | 40130459 | ||