siRNA Details
| Gene Symbol: Olfr1417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146936.1 | NP_667147.1 | 1 | 221 | ||
| GenBank mRNA | BC148189.1 | - | 1 | 221 | 40051854 | |
| BC153816.1 | - | 1 | 221 | 40043087 | ||
| Gene Symbol: Olfr1418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC150977.1 | - | 1 | 271 | 9087890 | |

Olfr1417
Genomic Map