siRNA Details
| Gene Symbol: Olfr1417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146936.1 | NP_667147.1 | 1 | 844 | ||
| GenBank mRNA | BC148189.1 | - | 1 | 844 | 40051854 | |
| BC153816.1 | - | 1 | 844 | 40043087 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1417| Gene Symbol: Olfr1417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146936.1 | NP_667147.1 | 1 | 844 | ||
| GenBank mRNA | BC148189.1 | - | 1 | 844 | 40051854 | |
| BC153816.1 | - | 1 | 844 | 40043087 | ||