siRNA Details
| Gene Symbol: Olfr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146937.1 | NP_667148.1 | 1 | 276 | ||
| GenBank mRNA | BC127997.1 | - | 1 | 280 | 40135782 | |
More Information
Other Species Targeted
Set Membership: |
|

Olfr63
Genomic Map