siRNA Details
| Gene Symbol: Olfr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146937.1 | NP_667148.1 | 1 | 772 | ||
| GenBank mRNA | BC127997.1 | - | 1 | 776 | 40135782 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr63| Gene Symbol: Olfr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146937.1 | NP_667148.1 | 1 | 772 | ||
| GenBank mRNA | BC127997.1 | - | 1 | 776 | 40135782 | |
Set Membership: |
|