siRNA Details
| Gene Symbol: Olfr354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146939.1 | NP_667150.1 | 1 | 269 | ||
| GenBank mRNA | BC119279.1 | - | 1 | 306 | 8734028 | |
| BC119283.1 | - | 1 | 306 | 8734032 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr354| Gene Symbol: Olfr354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146939.1 | NP_667150.1 | 1 | 269 | ||
| GenBank mRNA | BC119279.1 | - | 1 | 306 | 8734028 | |
| BC119283.1 | - | 1 | 306 | 8734032 | ||