siRNA Details
| Gene Symbol: Olfr351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146942.1 | NP_667153.1 | 1 | 239 | ||
| Gene Symbol: Olfr353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146941.1 | NP_667152.1 | 1 | 239 | ||

Olfr351
Genomic Map