siRNA Details
| Gene Symbol: Olfr342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146948.1 | NP_667159.1 | 1 | 498 | ||
| GenBank mRNA | BC120521.1 | - | 1 | 524 | 40129444 | |
| BC120523.1 | - | 1 | 524 | 40129446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr342| Gene Symbol: Olfr342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146948.1 | NP_667159.1 | 1 | 498 | ||
| GenBank mRNA | BC120521.1 | - | 1 | 524 | 40129444 | |
| BC120523.1 | - | 1 | 524 | 40129446 | ||