siRNA Details
| Gene Symbol: Olfr341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146950.1 | NP_667161.1 | 1 | 595 | ||
| GenBank mRNA | BC130244.1 | - | 1 | 599 | 40142305 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr341| Gene Symbol: Olfr341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146950.1 | NP_667161.1 | 1 | 595 | ||
| GenBank mRNA | BC130244.1 | - | 1 | 599 | 40142305 | |