siRNA Details
| Gene Symbol: Olfr535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146954.1 | NP_667165.1 | 1 | 264 | ||
| GenBank mRNA | BC120826.1 | - | 1 | 287 | 40129749 | |
| BC120828.1 | - | 1 | 287 | 40129751 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr535| Gene Symbol: Olfr535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146954.1 | NP_667165.1 | 1 | 264 | ||
| GenBank mRNA | BC120826.1 | - | 1 | 287 | 40129749 | |
| BC120828.1 | - | 1 | 287 | 40129751 | ||