siRNA Details
| Gene Symbol: Olfr170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146957.1 | NP_667168.1 | 1 | 742 | ||
| GenBank mRNA | BC132585.1 | - | 1 | 779 | 40130862 | |
| BC132587.1 | - | 1 | 779 | 40130864 | ||
| Gene Symbol: Olfr171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146958.2 | NP_667169.2 | 1 | 745 | ||

Olfr170
Genomic Map