siRNA Details
Gene Symbol: Olfr631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271020.1 | NP_001257949.1 | 4 | 602 | ||
NM_146959.2 | NP_667170.2 | 5 | 541 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271020.1 | NP_001257949.1 | 4 | 602 | ||
NM_146959.2 | NP_667170.2 | 5 | 541 |