siRNA Details
| Gene Symbol: Olfr53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146960.2 | NP_667171.1 | 2 | 795 | ||
| GenBank mRNA | AF102529.1 | - | 377 | |||
| BC119325.1 | - | 1 | 595 | 8734074 | ||
| BC120694.1 | - | 1 | 597 | 40129617 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr53| Gene Symbol: Olfr53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146960.2 | NP_667171.1 | 2 | 795 | ||
| GenBank mRNA | AF102529.1 | - | 377 | |||
| BC119325.1 | - | 1 | 595 | 8734074 | ||
| BC120694.1 | - | 1 | 597 | 40129617 | ||