siRNA Details
Gene Symbol: Olfr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146960.2 | NP_667171.1 | 2 | 1063 | ||
GenBank mRNA | BC119325.1 | - | 1 | 863 | 8734074 | |
BC120694.1 | - | 1 | 865 | 40129617 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146960.2 | NP_667171.1 | 2 | 1063 | ||
GenBank mRNA | BC119325.1 | - | 1 | 863 | 8734074 | |
BC120694.1 | - | 1 | 865 | 40129617 |