siRNA Details
| Gene Symbol: Olfr541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146962.1 | NP_667173.1 | 1 | 593 | ||
| GenBank mRNA | BC119150.1 | - | 1 | 681 | 8733899 | |
| BC120836.1 | - | 1 | 681 | 40129759 | ||
| Gene Symbol: Olfr53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146960.2 | NP_667171.1 | 2 | 816 | ||
| GenBank mRNA | AF102529.1 | - | 398 | |||
| BC119325.1 | - | 1 | 616 | 8734074 | ||
| BC120694.1 | - | 1 | 618 | 40129617 | ||

Olfr541
Genomic Map