siRNA Details
| Gene Symbol: Olfr541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146962.1 | NP_667173.1 | 1 | 139 | ||
| GenBank mRNA | BC119150.1 | - | 1 | 227 | 8733899 | |
| BC120836.1 | - | 1 | 227 | 40129759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr541| Gene Symbol: Olfr541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146962.1 | NP_667173.1 | 1 | 139 | ||
| GenBank mRNA | BC119150.1 | - | 1 | 227 | 8733899 | |
| BC120836.1 | - | 1 | 227 | 40129759 | ||