siRNA Details
| Gene Symbol: Olfr1242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146968.2 | NP_667179.1 | 1 | 245 | ||
| GenBank mRNA | BC120865.1 | - | 1 | 245 | 40129788 | |
| BC125386.1 | - | 1 | 245 | 40129901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1242| Gene Symbol: Olfr1242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146968.2 | NP_667179.1 | 1 | 245 | ||
| GenBank mRNA | BC120865.1 | - | 1 | 245 | 40129788 | |
| BC125386.1 | - | 1 | 245 | 40129901 | ||