siRNA Details
| Gene Symbol: Olfr1243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146969.1 | NP_667180.1 | 1 | 238 | ||
| GenBank mRNA | BC111812.1 | - | 1 | 233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1243| Gene Symbol: Olfr1243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146969.1 | NP_667180.1 | 1 | 238 | ||
| GenBank mRNA | BC111812.1 | - | 1 | 233 | ||