siRNA Details
| Gene Symbol: Olfr1228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146971.1 | NP_667182.1 | 1 | 52 | ||
| GenBank mRNA | BC125451.1 | - | 1 | 135 | 40129966 | |
| BC125453.1 | - | 1 | 135 | 40129968 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1228| Gene Symbol: Olfr1228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146971.1 | NP_667182.1 | 1 | 52 | ||
| GenBank mRNA | BC125451.1 | - | 1 | 135 | 40129966 | |
| BC125453.1 | - | 1 | 135 | 40129968 | ||