siRNA Details
| Gene Symbol: Olfr1273-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146975.1 | NP_667186.1 | 1 | 566 | ||
| GenBank mRNA | BC104266.1 | - | 1 | 566 | 40040402 | |
| BC104267.1 | - | 1 | 566 | 40040403 | ||
| BC126917.1 | - | 1 | 566 | 40040394 | ||

Olfr1273-ps
Genomic Map