siRNA Details
| Gene Symbol: Olfr1257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146982.1 | NP_667193.1 | 1 | 107 | ||
| GenBank mRNA | BC137779.1 | - | 1 | 141 | 8860798 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1257| Gene Symbol: Olfr1257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146982.1 | NP_667193.1 | 1 | 107 | ||
| GenBank mRNA | BC137779.1 | - | 1 | 141 | 8860798 | |