siRNA Details
| Gene Symbol: Olfr1270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146985.2 | NP_667196.1 | 1 | 507 | ||
| GenBank mRNA | BC119113.1 | - | 1 | 507 | 8733862 | |
| BC119115.1 | - | 1 | 507 | 8733864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1270| Gene Symbol: Olfr1270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146985.2 | NP_667196.1 | 1 | 507 | ||
| GenBank mRNA | BC119113.1 | - | 1 | 507 | 8733862 | |
| BC119115.1 | - | 1 | 507 | 8733864 | ||