siRNA Details
Gene Symbol: Olfr457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146987.1 | NP_667198.1 | 1 | 820 | ||
GenBank mRNA | BC119371.1 | - | 1 | 844 | 8734120 | |
BC119373.1 | - | 1 | 844 | 8734122 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146987.1 | NP_667198.1 | 1 | 820 | ||
GenBank mRNA | BC119371.1 | - | 1 | 844 | 8734120 | |
BC119373.1 | - | 1 | 844 | 8734122 |