siRNA Details
| Gene Symbol: Olfr457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146987.1 | NP_667198.1 | 1 | 136 | ||
| GenBank mRNA | BC119371.1 | - | 1 | 160 | 8734120 | |
| BC119373.1 | - | 1 | 160 | 8734122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr457| Gene Symbol: Olfr457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146987.1 | NP_667198.1 | 1 | 136 | ||
| GenBank mRNA | BC119371.1 | - | 1 | 160 | 8734120 | |
| BC119373.1 | - | 1 | 160 | 8734122 | ||