siRNA Details
| Gene Symbol: PSMA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002792.3 | NP_002783.1 | 5 | 711 | ||
| GenBank mRNA | AF022815.1 | - | 5 | 580 | ||
| AF054185.1 | - | 5 | 641 | |||
| AK127210.1 | - | 2 | 2406 | |||
| AK312025.1 | - | 5 | 606 | |||
| BC001895.1 | - | 5 | 644 | |||
| BC004427.1 | - | 5 | 629 | 2819923 | ||
| BC014999.1 | - | 5 | 639 | |||
| BG575009.1 | - | 5 | 701 | |||
| BT007165.1 | - | 5 | 556 | |||
| GQ472224.1 | - | 5 | 629 | |||

PSMA7
Genomic Map