siRNA Details
| Gene Symbol: Olfr1496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146989.2 | NP_667200.2 | 1 | 338 | ||
| GenBank mRNA | BC137878.1 | - | 1 | 338 | 8860897 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1496| Gene Symbol: Olfr1496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146989.2 | NP_667200.2 | 1 | 338 | ||
| GenBank mRNA | BC137878.1 | - | 1 | 338 | 8860897 | |