siRNA Details
| Gene Symbol: Olfr201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146994.2 | NP_667205.2 | 1 | 517 | ||
| Gene Symbol: Olfr199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207550.2 | NP_997433.2 | 1 | 517 | ||
| GenBank mRNA | BC128028.1 | - | 1 | 521 | 40136725 | |

Olfr201
Genomic Map