siRNA Details
Gene Symbol: Olfr178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146997.2 | NP_667208.2 | 1 | 883 | ||
GenBank mRNA | BC111099.1 | - | 1 | 882 | ||
BC127221.1 | - | 1 | 881 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146997.2 | NP_667208.2 | 1 | 883 | ||
GenBank mRNA | BC111099.1 | - | 1 | 882 | ||
BC127221.1 | - | 1 | 881 |