siRNA Details
| Gene Symbol: Olfr181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146999.2 | NP_667210.2 | 2 | 1164 | ||
| GenBank mRNA | BC125397.1 | - | 1 | 877 | 40129912 | |
| BC131916.1 | - | 1 | 877 | 40130193 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr181| Gene Symbol: Olfr181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146999.2 | NP_667210.2 | 2 | 1164 | ||
| GenBank mRNA | BC125397.1 | - | 1 | 877 | 40129912 | |
| BC131916.1 | - | 1 | 877 | 40130193 | ||