siRNA Details
| Gene Symbol: Olfr173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147000.2 | NP_667211.2 | 1 | 692 | ||
| GenBank mRNA | BC120572.1 | - | 1 | 692 | 40129495 | |
| BC137837.1 | - | 1 | 692 | 8860856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr173| Gene Symbol: Olfr173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147000.2 | NP_667211.2 | 1 | 692 | ||
| GenBank mRNA | BC120572.1 | - | 1 | 692 | 40129495 | |
| BC137837.1 | - | 1 | 692 | 8860856 | ||